Please login to your online account to display your discounted pricing

anti-Synuclein-alpha (Nitrate Tyr125, Nitrate Tyr133), Clone: 24.8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP126380SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP126380SS

  • / Each
 

Description & Specifications

Specifications

Antigen Nitrate Tyr133)
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1, κ
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6622
Concentration 1mg/mL
Primary or Secondary Primary
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Regulatory Status RUO
Clone 24.8
Format Purified
Immunogen Nitrated human alpha-Synuclein [UniProt No. P37840]
Quantity 0.025mL
Gene Accession No. P37840