Please login to your online account to display your discounted pricing

anti-Synuclein-alpha, Clone: 3H9, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP192694

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP192694

  • / Each
 

Description & Specifications

Specifications

Immunogen Amino acids 61-95 of human alpha-Synuclein, which correspond to the “Non-Amyloid beta Component of Alzheimer's disease amyloid” (NAC). [UniProt No. P37840]
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blotting
Regulatory Status RUO
Host Species Mouse
Isotype IgG1
Antigen Synuclein-alpha
Quantity 0.1mL
Format Ascites
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Gene Accession No. P37840
Primary or Secondary Primary
Gene ID (Entrez) 6622
Test Specificity This antibody recognizes full length human and rodent alpha Synuclein. It will also bind human alpha Synuclein containing the A30P and A53T mutations.
Clone 3H9