Please login to your online account to display your discounted pricing

anti-Synuclein-alpha, Clone: 3H9, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 3H9
Host Species Murine
Isotype IgG1
Immunogen Amino acids 61-95 of human alpha-Synuclein, which correspond to the “Non-Amyloid beta Component of Alzheimer's disease amyloid” (NAC). [UniProt No. P37840]
Antigen Synuclein-alpha
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP192694

 
novus biologicals canada
NBP192694
Unlabeled 0.1mL Each for $329.00

NBP192694SS

 
novus biologicals canada
NBP192694SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone 3H9
Host Species Murine
Isotype IgG1
Immunogen Amino acids 61-95 of human alpha-Synuclein, which correspond to the “Non-Amyloid beta Component of Alzheimer's disease amyloid” (NAC). [UniProt No. P37840]
Antigen Synuclein-alpha
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Simple Western
Applications Western Blot
Test Specificity This antibody recognizes full length human and rodent alpha Synuclein. It will also bind human alpha Synuclein containing the A30P and A53T mutations.
Gene Accession No. P37840
Gene ID (Entrez) 6622