Please login to your online account to display your discounted pricing

anti-Syndecan-1/CD138, Clone: B-A38, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10064980SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10064980T

  • / Each
 

Description & Specifications

Specifications

Clone B-A38
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Isotype IgG1
Host Species Mouse
Antigen Syndecan-1/CD138
Format Purified
Gene ID (Entrez) 6382
Test Specificity NB100-64980 recognizes a heparan sulfate rich membrane glycoprotein of 200kD known as Syndecan-1 (CD138). Syndecan functions as a receptor for collagen, fibronectin and thrombospondin. NB100-64980 stains all plasma cells and plasma cell lines. No staining of other leucocytes is seen in peripheral blood or bone marrow, although some ALL cell lines express the antigen weakly. Some staining of epithelial and endothelial cells may be seen in immunohistology.
Formulation PBS, 1% BSA
Gene Accession No. P18827
Concentration 0.1mg/mL
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Quantity 0.02mg
Immunogen U266 cell line
Cross Reactivity Human
Regulatory Status RUO
Primary or Secondary Primary