Please login to your online account to display your discounted pricing

anti-Syndecan-1/CD138, Clone: B-A38, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10064980

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10064980

  • / Each
 

Description & Specifications

Specifications

Antigen Syndecan-1/CD138
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone B-A38
Concentration 0.1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS, 1% BSA
Gene Accession No. P18827
Host Species Murine
Immunogen U266 cell line
Isotype IgG1
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6382
Test Specificity NB100-64980 recognizes a heparan sulfate rich membrane glycoprotein of 200kD known as Syndecan-1 (CD138). Syndecan functions as a receptor for collagen, fibronectin and thrombospondin. NB100-64980 stains all plasma cells and plasma cell lines. No staining of other leucocytes is seen in peripheral blood or bone marrow, although some ALL cell lines express the antigen weakly. Some staining of epithelial and endothelial cells may be seen in immunohistology.
Target Species Human