Please login to your online account to display your discounted pricing

anti-Syndecan-1/CD138, Clone: B-A38, Novus Biologicals™

$142.00 - $442.22

Specifications

Clone B-A38
Host Species Mouse
Isotype IgG1
Immunogen U266 cell line
Antigen Syndecan-1/CD138
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10064980

 
novus biologicals canada
NB10064980
Unlabeled 0.1mg Each for $442.22

NB10064980T

 
novus biologicals canada
NB10064980SS
Unlabeled 0.02mg Each for $142.00
Description & Specifications

Specifications

Clone B-A38
Host Species Mouse
Isotype IgG1
Immunogen U266 cell line
Antigen Syndecan-1/CD138
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity NB100-64980 recognizes a heparan sulfate rich membrane glycoprotein of 200kD known as Syndecan-1 (CD138). Syndecan functions as a receptor for collagen, fibronectin and thrombospondin. NB100-64980 stains all plasma cells and plasma cell lines. No staining of other leucocytes is seen in peripheral blood or bone marrow, although some ALL cell lines express the antigen weakly. Some staining of epithelial and endothelial cells may be seen in immunohistology.
Gene Accession No. P18827
Gene ID (Entrez) 6382
Formulation PBS, 1% BSA
Concentration 0.1mg/mL