Please login to your online account to display your discounted pricing

anti-Survivin, Clone: 8E2, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 8E2
Host Species Mouse
Isotype IgG1, κ
Immunogen Full length recombinant human Survivin [UniProt No. O15392]
Antigen Survivin
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB500644

 
novus biologicals canada
NB500644
Unlabeled 0.1mL Each for $309.05

NB500644SS

 
novus biologicals canada
NB500644SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 8E2
Host Species Mouse
Isotype IgG1, κ
Immunogen Full length recombinant human Survivin [UniProt No. O15392]
Antigen Survivin
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications In vitro Assay
Applications Neutralization
Applications Western Blotting
Gene Accession No. O15392
Gene ID (Entrez) 332
Formulation Tris-glycine, 150mM NaCl
Concentration 1mg/mL