Please login to your online account to display your discounted pricing

anti-SUMO1, Clone: SM1/495, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone SM1/495
Host Species Mouse
Isotype κ
Immunogen Recombinant human SUMO1 protein
Antigen SUMO1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232901

 
novus biologicals canada
NBP232901
Unlabeled 0.5mL Each for $321.00

NBP2329011M

 
novus biologicals canada
NBP2329011ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone SM1/495
Host Species Mouse
Isotype κ
Immunogen Recombinant human SUMO1 protein
Antigen SUMO1
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This MAb is specific to SUMO-1 and shows no cross-reaction with either SUMO-2 or SUMO-3. The small ubiquitin-related modifier (SUMO) proteins, which include SUMO-1, SUMO-2 and SUMO-3, belong to the ubiquitin-like protein family. Like ubiquitin, the SUMO proteins are synthesized as precursor proteins that undergo processing before conjugation to target proteins. Also, both utilize the E1, E2, and E3 cascade enzymes for conjugation. However, SUMO and ubiquitin differ with respect to targeting. Ubiquitination predominantly targets proteins for degradation, whereas sumoylation targets proteins to a variety of cellular processing, including nuclear transport, transcriptional regulation, apoptosis and protein stability. The unconjugated SUMO-1 protein localizes to the nuclear membrane. Shows broad species reactivity.
Gene Accession No. P63165
Gene ID (Entrez) 7341
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL