Please login to your online account to display your discounted pricing

anti-STAT6, Clone: 177C322.1, Novus Biologicals™

$109.00 - $307.00

Specifications

Clone 177C322.1
Host Species Mouse
Isotype IgG1
Immunogen A portion of amino acids 600-650 of human STAT6 was used as the immunogen for the antibody.
Antigen STAT6
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225241

 
novus biologicals canada
NBP225241
Unlabeled 0.1mg Each for $307.00

NBP225241SS

 
novus biologicals canada
NBP225241SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Clone 177C322.1
Host Species Mouse
Isotype IgG1
Immunogen A portion of amino acids 600-650 of human STAT6 was used as the immunogen for the antibody.
Antigen STAT6
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity This product reacts with Human and Mouse.
Gene Accession No. NP_003144
Gene ID (Entrez) 6778
Formulation 0.1mg STAT6 antibody in 0.2mL PBS containing 0.05% BSA
Concentration 0.5mg/mL