Please login to your online account to display your discounted pricing

anti-ST6 Gal Sialyltransferase 1/ST6GAL1/CD75, Clone: LN-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP229466

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229466

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 0.5mL
Immunogen 1984).
Clone LN-1
Test Specificity Recognizes a neuraminidase-sensitive sialoprotein (CDw75), present on cell membrane and cytoplasm of germinal center B-cells and derived lymphomas. This MAb reacts with RBC precursors of bone marrow, ductal and ciliated epithelial cells of kidney, breast, prostate, pancreas, lung, and with glioblastomas, astrocytomas, and Reed Sternberg cells in lymphocyte predominant Hodgkin's disease.
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Formulation Culture Medium
Gene ID (Entrez) 6480
Gene Accession No. P15907
Primary or Secondary Primary
Host Species Mouse
Cross Reactivity Human
Format Concentrated
Antigen ST6 Gal Sialyltransferase 1/ST6GAL1/CD75
Isotype κ