Please login to your online account to display your discounted pricing

anti-ST6 Gal Sialyltransferase 1/ST6GAL1/CD75, Clone: LN-1, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone LN-1
Host Species Mouse
Isotype κ
Immunogen 1984).
Antigen ST6 Gal Sialyltransferase 1/ST6GAL1/CD75
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP229466

 
novus biologicals canada
NBP229466
Unlabeled 0.5mL Each for $321.00

NBP2294661M

 
novus biologicals canada
NBP2294661ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone LN-1
Host Species Mouse
Isotype κ
Immunogen 1984).
Antigen ST6 Gal Sialyltransferase 1/ST6GAL1/CD75
Monoclonal or Polyclonal Monoclonal
Format Concentrated
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Test Specificity Recognizes a neuraminidase-sensitive sialoprotein (CDw75), present on cell membrane and cytoplasm of germinal center B-cells and derived lymphomas. This MAb reacts with RBC precursors of bone marrow, ductal and ciliated epithelial cells of kidney, breast, prostate, pancreas, lung, and with glioblastomas, astrocytomas, and Reed Sternberg cells in lymphocyte predominant Hodgkin's disease.
Gene Accession No. P15907
Gene ID (Entrez) 6480
Formulation Culture Medium