Please login to your online account to display your discounted pricing

anti-ST6 Gal Sialyltransferase 1/ST6GAL1/CD75, Clone: LN-1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2294661ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2294661M

  • / Each
 

Description & Specifications

Specifications

Test Specificity Recognizes a neuraminidase-sensitive sialoprotein (CDw75), present on cell membrane and cytoplasm of germinal center B-cells and derived lymphomas. This MAb reacts with RBC precursors of bone marrow, ductal and ciliated epithelial cells of kidney, breast, prostate, pancreas, lung, and with glioblastomas, astrocytomas, and Reed Sternberg cells in lymphocyte predominant Hodgkin's disease.
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Host Species Mouse
Antigen ST6 Gal Sialyltransferase 1/ST6GAL1/CD75
Format Concentrated
Immunogen 1984).
Gene ID (Entrez) 6480
Primary or Secondary Primary
Gene Accession No. P15907
Conjugate Unlabeled
Formulation Culture Medium
Monoclonal or Polyclonal Monoclonal
Quantity 1mL
Isotype κ
Cross Reactivity Human
Clone LN-1