Please login to your online account to display your discounted pricing

anti-Smallpox B5R, Clone: 1J2F3, Novus Biologicals™

$139.00 - $259.02

Specifications

Clone 1J2F3
Host Species Mouse
Isotype κ
Immunogen Within the range of Amino acids 33-62 of the B5R protein were used as the immunogen.
Antigen Smallpox B5R
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224699SS

 
novus biologicals canada
NBP224699SS
Unlabeled 0.025mg Each for $139.00

NBP224699

 
novus biologicals canada
NBP224699
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Clone 1J2F3
Host Species Mouse
Isotype κ
Immunogen Within the range of Amino acids 33-62 of the B5R protein were used as the immunogen.
Antigen Smallpox B5R
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen for this phospho antibody is 100% homologous in Human, Chimpanzee, Dog, Rat, Mouse, Chicken and Zebrafish and 92% homologous in Xenopus.
Gene Accession No. P33823
Gene ID (Entrez) 1486562
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL