Please login to your online account to display your discounted pricing

anti-SM047, Clone: SM047, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP197921

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP197921

  • / Each
 

Description & Specifications

Specifications

Isotype IgM
Quantity 0.1mL
Immunogen Cell membranes from human ovarian serous cystadenocarcinmoma and cells from human ovarian cancer cell line UWOV1.
Cross Reactivity Human
Format Ascites
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Antigen SM047
Clone SM047
Monoclonal or Polyclonal Monoclonal
Test Specificity Glycocalyx of human ovarian carcinoma cells.
Primary or Secondary Primary
Conjugate Unlabeled