Please login to your online account to display your discounted pricing

anti-SLC26A7, Clone: 14H5, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11060965

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11060965

  • / Each
 

Description & Specifications

Specifications

Isotype IgG2a
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Immunogen A fusion protein containing the C-terminal region of mouse SLC26A7. [Swiss-Prot No. Q24JS8]
Antigen SLC26A7
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 115111
Format Purified
Concentration 1.7mg/mL
Quantity 0.1mL
Host Species Mouse
Cross Reactivity Mouse
Clone 14H5
Regulatory Status RUO
Conjugate Unlabeled
Formulation Tris-glycine, 150mM NaCl
Primary or Secondary Primary