Please login to your online account to display your discounted pricing

anti-SLC26A7, Clone: 14H5, Novus Biologicals™

$109.00 - $309.05

Specifications

Clone 14H5
Host Species Mouse
Isotype IgG2a
Immunogen A fusion protein containing the C-terminal region of mouse SLC26A7. [Swiss-Prot No. Q24JS8]
Antigen SLC26A7
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11060965

 
novus biologicals canada
NB11060965
Unlabeled 0.1mL Each for $309.05

NB11060965T

 
novus biologicals canada
NB11060965SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Clone 14H5
Host Species Mouse
Isotype IgG2a
Immunogen A fusion protein containing the C-terminal region of mouse SLC26A7. [Swiss-Prot No. Q24JS8]
Antigen SLC26A7
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Gene ID (Entrez) 115111
Formulation Tris-glycine, 150mM NaCl
Concentration 1.7mg/mL