Please login to your online account to display your discounted pricing

anti-SLC26A7, Clone: 14H5, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11060965SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11060965T

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Primary or Secondary Primary
Regulatory Status RUO
Antigen SLC26A7
Conjugate Unlabeled
Cross Reactivity Mouse
Quantity 0.025mL
Clone 14H5
Immunogen A fusion protein containing the C-terminal region of mouse SLC26A7. [Swiss-Prot No. Q24JS8]
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Isotype IgG2a
Formulation Tris-glycine, 150mM NaCl
Concentration 1.7mg/mL
Gene ID (Entrez) 115111
Monoclonal or Polyclonal Monoclonal