Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, Clone: WM53, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP2329311ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP2329311M

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Isotype κ
Host Species Mouse
Antigen Siglec-3/CD33
Format Purified
Conjugate Unlabeled
Gene ID (Entrez) 945
Concentration 0.2mg/mL
Primary or Secondary Primary
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P20138
Quantity 1mL
Regulatory Status RUO
Clone WM53
Immunogen Human AML cells