Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, Clone: C33/69, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232820

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232820

  • / Each
 

Description & Specifications

Specifications

Immunogen Recombinant human CD33 protein
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Quantity 0.5mL
Cross Reactivity Human
Clone C33/69
Isotype κ
Formulation PBS with 0.05% BSA.
Primary or Secondary Primary
Concentration 0.2mg/mL
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Gene Accession No. P20138
Gene ID (Entrez) 945
Regulatory Status RUO
Host Species Mouse
Antigen Siglec-3/CD33
Format Purified