Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, Clone: C33/69, Novus Biologicals™

$321.00 - $429.00

Specifications

Clone C33/69
Host Species Mouse
Isotype κ
Immunogen Recombinant human CD33 protein
Antigen Siglec-3/CD33
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP232820

 
novus biologicals canada
NBP232820
Unlabeled 0.5mL Each for $321.00

NBP2328201M

 
novus biologicals canada
NBP2328201ML
Unlabeled 1mL Each for $429.00
Description & Specifications

Specifications

Clone C33/69
Host Species Mouse
Isotype κ
Immunogen Recombinant human CD33 protein
Antigen Siglec-3/CD33
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Gene Accession No. P20138
Gene ID (Entrez) 945
Formulation PBS with 0.05% BSA.
Concentration 0.2mg/mL