Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, Clone: C33/69, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP2328201ML

 View all options for this product


For Research Use Only

Catalog No. NBP2328201M


Description & Specifications


Antigen Siglec-3/CD33
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Clone C33/69
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P20138
Host Species Mouse
Immunogen Recombinant human CD33 protein
Isotype κ
Quantity 1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 945