Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, Clone: C33/68, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP232819

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP232819

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Antigen Siglec-3/CD33
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Format Purified
Isotype κ
Quantity 0.5mL
Primary or Secondary Primary
Gene ID (Entrez) 945
Clone C33/68
Concentration 0.2mg/mL
Conjugate Unlabeled
Gene Accession No. P20138
Monoclonal or Polyclonal Monoclonal
Formulation PBS with 0.05% BSA.
Immunogen Recombinant human CD33 protein
Cross Reactivity Human