Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, Clone: 44M12D3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222377

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222377

  • / Each
 

Description & Specifications

Specifications

Quantity 0.1mg
Antigen Siglec-3/CD33
Host Species Mouse
Format Purified
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Cross Reactivity Human
Immunogen Amino acids 42-159 of human CD33 were used as the immunogen for the antibody.
Clone 44M12D3
Gene ID (Entrez) 945
Monoclonal or Polyclonal Monoclonal
Concentration 0.5mg/mL
Primary or Secondary Primary
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Conjugate Unlabeled
Gene Accession No. AAH28152
Isotype κ