Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, Clone: 44M12D3, Novus Biologicals™

$109.00 - $329.00

Specifications

Clone 44M12D3
Host Species Murine
Isotype κ
Immunogen Amino acids 42-159 of human CD33 were used as the immunogen for the antibody.
Antigen Siglec-3/CD33
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222377SS

 
novus biologicals canada
NBP222377SS
Unlabeled 0.025mg Each for $109.00

NBP222377

 
novus biologicals canada
NBP222377
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Clone 44M12D3
Host Species Murine
Isotype κ
Immunogen Amino acids 42-159 of human CD33 were used as the immunogen for the antibody.
Antigen Siglec-3/CD33
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. AAH28152
Gene ID (Entrez) 945
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Concentration 0.5mg/mL