Please login to your online account to display your discounted pricing

anti-Siglec-3/CD33, Clone: 44M12D3, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222377SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222377SS

  • / Each
 

Description & Specifications

Specifications

Antigen Siglec-3/CD33
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Clone 44M12D3
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Gene Accession No. AAH28152
Host Species Murine
Immunogen Amino acids 42-159 of human CD33 were used as the immunogen for the antibody.
Isotype κ
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 945
Target Species Human