Please login to your online account to display your discounted pricing

anti-SIAH2, Clone: 24E6H3, Novus Biologicals™

$139.00 - $329.00

Specifications

Clone 24E6H3
Host Species Murine
Isotype IgG1
Immunogen Synthetic peptide made to an N-terminal portion of the drosophila SINA protein (within residues 1-100). [Swiss-Prot No. P21461]
Antigen SIAH2
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11088113T

 
novus biologicals canada
NB11088113SS
Unlabeled 0.125mL Each for $139.00

NB11088113

 
novus biologicals canada
NB11088113
Unlabeled 0.5mL Each for $329.00
Description & Specifications

Specifications

Clone 24E6H3
Host Species Murine
Isotype IgG1
Immunogen Synthetic peptide made to an N-terminal portion of the drosophila SINA protein (within residues 1-100). [Swiss-Prot No. P21461]
Antigen SIAH2
Monoclonal or Polyclonal Monoclonal
Format Supernatant
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This is specific for human SIAH2 and Drosophila SINA in proliferating cells and stem cells. This does not detect Drosophila SINAH or normal, nondividing cells. Human, Porcine, Drosophila. Does not react with mouse.
Gene ID (Entrez) 6478