Please login to your online account to display your discounted pricing

anti-SH3PX1, Clone: 1B5, Novus Biologicals™

$494.00 - $563.15

Specifications

Clone 1B5
Host Species Murine
Isotype IgG1
Immunogen Full length human recombinant protein of human SNX9(NP_057308) produced in HEK293T cell.
Antigen SH3PX1
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP202609SS

 
novus biologicals canada
NBP202609SS
Unlabeled 0.02mL Each for $563.15

NBP202609

 
novus biologicals canada
NBP202609
Unlabeled 0.1mL Each for $494.00
Description & Specifications

Specifications

Clone 1B5
Host Species Murine
Isotype IgG1
Immunogen Full length human recombinant protein of human SNX9(NP_057308) produced in HEK293T cell.
Antigen SH3PX1
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene ID (Entrez) 51429
Formulation PBS (pH 7.3) containing 1% BSA, 50% glycerol