Please login to your online account to display your discounted pricing

anti-SH3PX1, Clone: 1B5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP202609SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP202609SS

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Full length human recombinant protein of human SNX9(NP_057308) produced in HEK293T cell.
Regulatory Status RUO
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Primate
Cross Reactivity Rat
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Formulation PBS (pH 7.3) containing 1% BSA, 50% glycerol
Conjugate Unlabeled
Gene ID (Entrez) 51429
Antigen SH3PX1
Format Affinity Purified
Isotype IgG1
Quantity 0.02mL
Clone 1B5