Please login to your online account to display your discounted pricing

anti-Separase, Clone: XJ11-4D7, Novus Biologicals™

$139.00 - $309.05

Specifications

Clone XJ11-4D7
Host Species Mouse
Isotype IgG2a
Immunogen Maltose-Binding Protein fusion of C-terminal fragment of human Separase (amino acids 1866-1996). [UniProt No. Q14674]
Antigen Separase
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100330

 
novus biologicals canada
NB100330
Unlabeled 0.1mL Each for $309.05

NB100330SS

 
novus biologicals canada
NB100330SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Clone XJ11-4D7
Host Species Mouse
Isotype IgG2a
Immunogen Maltose-Binding Protein fusion of C-terminal fragment of human Separase (amino acids 1866-1996). [UniProt No. Q14674]
Antigen Separase
Monoclonal or Polyclonal Monoclonal
Format Ascites
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Gene Accession No. Q14674
Gene ID (Entrez) 9700