Please login to your online account to display your discounted pricing

anti-Semaphorin 4D/CD100, Clone: A8, Novus Biologicals™

$109.00 - $252.00

Specifications

Clone A8
Host Species Murine
Isotype κ
Immunogen PHA-activated human lymphocytes were used as the immunogen for the CD100 antibody.
Antigen Semaphorin 4D/CD100
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225198SS

 
novus biologicals canada
NBP225198SS
Unlabeled 0.025mg Each for $109.00

NBP225198

 
novus biologicals canada
NBP225198
Unlabeled 0.1mg Each for $252.00
Description & Specifications

Specifications

Clone A8
Host Species Murine
Isotype κ
Immunogen PHA-activated human lymphocytes were used as the immunogen for the CD100 antibody.
Antigen Semaphorin 4D/CD100
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunoprecipitation
Test Specificity This product reacts with Human.
Gene Accession No. NP_006369
Gene ID (Entrez) 10507
Formulation CD100 antibody at 0.5mg/mL in PBS containing 0.05% BSA
Concentration 0.5mg/mL