Please login to your online account to display your discounted pricing

anti-Mouse SCF/c-kit Ligand, Polyclonal, R&D Systems

$458.10 - $458.10

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q78ED8
Gene ID (Entrez) 17311
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

AB455NA

 
r & d systems
RDSAB455NA
Unlabeled 1mg Each for $458.10

AF455NA

 
r & d systems
RDSAF455NA
Unlabeled 100μg Each for $458.10
Description & Specifications

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Gene Accession No. Q78ED8
Gene ID (Entrez) 17311

Host Species: Goat
Species Reactivity:Mouse
Immunogen:E. coli-derived recombinant mouse SCF/c-kit Ligand, Lys26-Ala189, Accession No. Q78ED8

Applications:

Neutralization, Western Blotting