Please login to your online account to display your discounted pricing

anti-Mouse SCF/c-kit Ligand, Polyclonal, R&D Systems


Manufacturer: r & d systems  RDSAF455NA

 View more versions of this product

Catalog No. AF455NA

  • / Each
 

Description & Specifications

Specifications

Antigen SCF/c-kit Ligand
Conjugate Unlabeled
Cross Reactivity Mouse
Format Affinity Purified
Formulation Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. &asterisk;Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS.
Gene Accession No. Q78ED8
Host Species Goat
Immunogen E. coli-derived recombinant mouse SCF/c-kit Ligand, Lys26-Ala189, Accession No. Q78ED8
Isotype IgG
Quantity 100μg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 17311
Target Species Murine

SCF/c-kit Ligand Polyclonal antibody AF455SP Specifically detects SCF/c-kit Ligand in Mouse samples. It is validated for ELISA Capture, Neutralization, Western Blotting