Please login to your online account to display your discounted pricing

anti-Sars Spike Protein, Clone: 17F706, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224942

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224942

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.2% gelatin
Test Specificity This antibody is reactive to Human.
Gene Accession No. P49591
Quantity 0.1mg
Cross Reactivity Human
Regulatory Status RUO
Isotype κ
Format Affinity Purified
Immunogen Amino acids 1124-1140 of human SARS-Spike protein were used as the immunogen
Antigen Sars Spike Protein
Applications Western Blotting
Clone 17F706
Concentration 0.5mg/mL
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Gene ID (Entrez) 6301
Conjugate Unlabeled