Please login to your online account to display your discounted pricing

anti-Sars Spike Protein, Clone: 17F706, Novus Biologicals™

$142.00 - $409.37

Specifications

Clone 17F706
Host Species Murine
Isotype κ
Immunogen Amino acids 1124-1140 of human SARS-Spike protein were used as the immunogen
Antigen Sars Spike Protein
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224942

 
novus biologicals canada
NBP224942
Unlabeled 0.1mg Each for $409.37

NBP224942SS

 
novus biologicals canada
NBP224942SS
Unlabeled 0.025mg Each for $142.00
Description & Specifications

Specifications

Clone 17F706
Host Species Murine
Isotype κ
Immunogen Amino acids 1124-1140 of human SARS-Spike protein were used as the immunogen
Antigen Sars Spike Protein
Monoclonal or Polyclonal Monoclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity This antibody is reactive to Human.
Gene Accession No. P49591
Gene ID (Entrez) 6301
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL