Please login to your online account to display your discounted pricing

anti-Sars Spike Protein, Clone: 17F706, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224942SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224942SS

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Isotype κ
Concentration 0.5mg/mL
Cross Reactivity Human
Quantity 0.025mg
Immunogen Amino acids 1124-1140 of human SARS-Spike protein were used as the immunogen
Regulatory Status RUO
Host Species Mouse
Antigen Sars Spike Protein
Format Affinity Purified
Clone 17F706
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Applications Western Blotting
Test Specificity This antibody is reactive to Human.
Gene ID (Entrez) 6301
Gene Accession No. P49591
Primary or Secondary Primary