Please login to your online account to display your discounted pricing

anti-Sars Spike Protein, Clone: 16F1071, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224746

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224746

  • / Each
 

Description & Specifications

Specifications

Antigen Sars Spike Protein
Applications Western Blot
Clone 16F1071
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Gene Accession No. P49591
Host Species Murine
Immunogen The antibody was developed using a synthetic peptide from the putative SARS Spike glycoprotein (Genbank accession no. NP_828851) corresponding to amino acids 19-45.
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6301
Test Specificity The C1.18.4 hybridoma cell line was grown and tissue culture supernatant was purified using affinity chromatography.
Target Species Human