Please login to your online account to display your discounted pricing

anti-Sars Spike Protein, Clone: 16F1071, Novus Biologicals™

$139.00 - $368.59

Specifications

Clone 16F1071
Host Species Murine
Isotype κ
Immunogen The antibody was developed using a synthetic peptide from the putative SARS Spike glycoprotein (Genbank accession no. NP_828851) corresponding to amino acids 19-45.
Antigen Sars Spike Protein
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224746

 
novus biologicals canada
NBP224746
Unlabeled 0.1mg Each for $368.59

NBP224746SS

 
novus biologicals canada
NBP224746SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 16F1071
Host Species Murine
Isotype κ
Immunogen The antibody was developed using a synthetic peptide from the putative SARS Spike glycoprotein (Genbank accession no. NP_828851) corresponding to amino acids 19-45.
Antigen Sars Spike Protein
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The C1.18.4 hybridoma cell line was grown and tissue culture supernatant was purified using affinity chromatography.
Gene Accession No. P49591
Gene ID (Entrez) 6301
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL