Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Clone: Ncap11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224747

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224747

  • / Each
 

Description & Specifications

Specifications

Formulation Antibody in 200μL PBS containing 0.2% gelatin
Isotype κ
Primary or Secondary Primary
Cross Reactivity Human
Immunogen The antibody was developed by immunizing mice with full-length recombinant SARS nucleocapsid protein (Genbank accession no. NP_828858). The specific epitope has not been determined.
Format Purified
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Quantity 0.2mL
Antigen SARS Nucleocapsid Protein
Clone Ncap11
Conjugate Unlabeled
Regulatory Status RUO
Gene Accession No. P49591
Gene ID (Entrez) 6301
Monoclonal or Polyclonal Monoclonal
Test Specificity This antibody is reactive to Human.