Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Clone: Ncap11, Novus Biologicals™

$142.00 - $409.37

Specifications

Clone Ncap11
Host Species Murine
Isotype κ
Immunogen The antibody was developed by immunizing mice with full-length recombinant SARS nucleocapsid protein (Genbank accession no. NP_828858). The specific epitope has not been determined.
Antigen SARS Nucleocapsid Protein
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224747SS

 
novus biologicals canada
NBP224747SS
Unlabeled 0.05mL Each for $142.00

NBP224747

 
novus biologicals canada
NBP224747
Unlabeled 0.2mL Each for $409.37
Description & Specifications

Specifications

Clone Ncap11
Host Species Murine
Isotype κ
Immunogen The antibody was developed by immunizing mice with full-length recombinant SARS nucleocapsid protein (Genbank accession no. NP_828858). The specific epitope has not been determined.
Antigen SARS Nucleocapsid Protein
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity This antibody is reactive to Human.
Gene Accession No. P49591
Gene ID (Entrez) 6301
Formulation Antibody in 200μL PBS containing 0.2% gelatin