Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Clone: Ncap11, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224747SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224747SS

  • / Each
 

Description & Specifications

Specifications

Antigen SARS Nucleocapsid Protein
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Clone Ncap11
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation Antibody in 200μL PBS containing 0.2% gelatin
Gene Accession No. P49591
Host Species Murine
Immunogen The antibody was developed by immunizing mice with full-length recombinant SARS nucleocapsid protein (Genbank accession no. NP_828858). The specific epitope has not been determined.
Isotype κ
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6301
Test Specificity This antibody is reactive to Human.
Target Species Human