Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Clone: 18F629.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224745

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224745

  • / Each
 

Description & Specifications

Specifications

Test Specificity The Cyclin D1 amino acid sequence used as immunogen for the antibody is 100% homologous in Human and Monkey, and 88% homologous in Mouse and Rat.
Cross Reactivity Human
Quantity 0.2mL
Immunogen The antibody was developed by immunizing mice with a KLH-conjugated peptide corresponding to amino acids 354-385 of human nucleocapsid protein (Genbank accession no. NP_828858).
Regulatory Status RUO
Format Purified
Host Species Mouse
Applications Western Blotting
Isotype IgM
Clone 18F629.1
Antigen SARS Nucleocapsid Protein
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6301
Gene Accession No. P49591
Primary or Secondary Primary
Conjugate Unlabeled
Formulation Antibody in 200μL PBS containing 0.2% gelatin