Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Clone: 18F629.1, Novus Biologicals™

$142.00 - $409.37

Specifications

Clone 18F629.1
Host Species Murine
Isotype IgM
Immunogen The antibody was developed by immunizing mice with a KLH-conjugated peptide corresponding to amino acids 354-385 of human nucleocapsid protein (Genbank accession no. NP_828858).
Antigen SARS Nucleocapsid Protein
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224745SS

 
novus biologicals canada
NBP224745SS
Unlabeled 0.05mL Each for $142.00

NBP224745

 
novus biologicals canada
NBP224745
Unlabeled 0.2mL Each for $409.37
Description & Specifications

Specifications

Clone 18F629.1
Host Species Murine
Isotype IgM
Immunogen The antibody was developed by immunizing mice with a KLH-conjugated peptide corresponding to amino acids 354-385 of human nucleocapsid protein (Genbank accession no. NP_828858).
Antigen SARS Nucleocapsid Protein
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The Cyclin D1 amino acid sequence used as immunogen for the antibody is 100% homologous in Human and Monkey, and 88% homologous in Mouse and Rat.
Gene Accession No. P49591
Gene ID (Entrez) 6301
Formulation Antibody in 200μL PBS containing 0.2% gelatin