Please login to your online account to display your discounted pricing

anti-SARS Nucleocapsid Protein, Clone: 18F629.1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224745SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224745SS

  • / Each
 

Description & Specifications

Specifications

Antigen SARS Nucleocapsid Protein
Applications Western Blot
Clone 18F629.1
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation Antibody in 200μL PBS containing 0.2% gelatin
Gene Accession No. P49591
Host Species Murine
Immunogen The antibody was developed by immunizing mice with a KLH-conjugated peptide corresponding to amino acids 354-385 of human nucleocapsid protein (Genbank accession no. NP_828858).
Isotype IgM
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6301
Test Specificity The Cyclin D1 amino acid sequence used as immunogen for the antibody is 100% homologous in Human and Monkey, and 88% homologous in Mouse and Rat.
Target Species Human