Please login to your online account to display your discounted pricing

anti-S100A8/A9, Clone: 48M7C7, Novus Biologicals™

$139.00 - $307.00

Specifications

Clone 48M7C7
Host Species Murine
Isotype κ
Immunogen Within the range of amino acids 2-105 of human S100A8 were used as the immunogen for the antibody.
Antigen S100A8/A9
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225256

 
novus biologicals canada
NBP225256
Unlabeled 0.1mg Each for $307.00

NBP225256SS

 
novus biologicals canada
NBP225256SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 48M7C7
Host Species Murine
Isotype κ
Immunogen Within the range of amino acids 2-105 of human S100A8 were used as the immunogen for the antibody.
Antigen S100A8/A9
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity This product reacts with Human and Mouse.
Gene ID (Entrez) 6279
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Concentration 0.5mg/mL