Please login to your online account to display your discounted pricing

anti-S100A8/A9, Clone: 48M5F4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225260SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225260SS

  • / Each
 

Description & Specifications

Specifications

Isotype κ
Concentration 0.5mg/mL
Quantity 0.025mg
Cross Reactivity Human
Immunogen Within the range of amino acids 2-101 of human S100A8 were used as the immunogen for the antibody.
Regulatory Status RUO
Format Purified
Host Species Mouse
Antigen S100A8/A9
Applications Western Blotting
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Primary or Secondary Primary
Clone 48M5F4
Conjugate Unlabeled
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6279
Test Specificity This product reacts with Human.