Please login to your online account to display your discounted pricing

anti-S100A8/A9, Clone: 48M5F4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP225260

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP225260

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Isotype κ
Quantity 0.1mg
Immunogen Within the range of amino acids 2-101 of human S100A8 were used as the immunogen for the antibody.
Regulatory Status RUO
Host Species Mouse
Format Purified
Applications Western Blotting
Antigen S100A8/A9
Clone 48M5F4
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 6279
Test Specificity This product reacts with Human.
Concentration 0.5mg/mL
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Conjugate Unlabeled