Please login to your online account to display your discounted pricing

anti-S100A8/A9, Clone: 48M5F4, Novus Biologicals™

$139.00 - $307.00

Specifications

Clone 48M5F4
Host Species Mouse
Isotype κ
Immunogen Within the range of amino acids 2-101 of human S100A8 were used as the immunogen for the antibody.
Antigen S100A8/A9
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP225260

 
novus biologicals canada
NBP225260
Unlabeled 0.1mg Each for $307.00

NBP225260SS

 
novus biologicals canada
NBP225260SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 48M5F4
Host Species Mouse
Isotype κ
Immunogen Within the range of amino acids 2-101 of human S100A8 were used as the immunogen for the antibody.
Antigen S100A8/A9
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity This product reacts with Human.
Gene ID (Entrez) 6279
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Concentration 0.5mg/mL