Please login to your online account to display your discounted pricing

anti-RUNX3/CBFA3, Clone: 2B9G4, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222390SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222390SS

  • / Each
 

Description & Specifications

Specifications

Conjugate Unlabeled
Immunogen Amino acids 126-310 of human Runx3 were used as the immunogen for this antibody.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Isotype κ
Regulatory Status RUO
Cross Reactivity Human
Quantity 0.025mg
Format Purified
Host Species Mouse
Antigen RUNX3/CBFA3
Clone 2B9G4
Gene Accession No. NP_001026850.1
Concentration 0.5mg/mL
Gene ID (Entrez) 864
Monoclonal or Polyclonal Monoclonal
Formulation 0.1mg antibody in 0.2mL PBS and 0.05% BSA