Please login to your online account to display your discounted pricing

anti-RUNX3/CBFA3, Clone: 2B10E8, Novus Biologicals™

$139.00 - $307.00

Specifications

Clone 2B10E8
Host Species Murine
Isotype κ
Immunogen Amino acids 126-310 of human Runx3 were used as the immunogen for this antibody.
Antigen RUNX3/CBFA3
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222391

 
novus biologicals canada
NBP222391
Unlabeled 0.1mg Each for $307.00

NBP222391SS

 
novus biologicals canada
NBP222391SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Clone 2B10E8
Host Species Murine
Isotype κ
Immunogen Amino acids 126-310 of human Runx3 were used as the immunogen for this antibody.
Antigen RUNX3/CBFA3
Monoclonal or Polyclonal Monoclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. NP_001026850.1
Gene ID (Entrez) 864
Formulation 0.1mg antibody in 0.2mL PBS and 0.05% BSA
Concentration 0.5mg/mL