Please login to your online account to display your discounted pricing

anti-RUNX3/CBFA3, Clone: 2B10E8, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222391SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222391SS

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Amino acids 126-310 of human Runx3 were used as the immunogen for this antibody.
Regulatory Status RUO
Isotype κ
Format Purified
Host Species Mouse
Quantity 0.025mg
Cross Reactivity Human
Antigen RUNX3/CBFA3
Clone 2B10E8
Formulation 0.1mg antibody in 0.2mL PBS and 0.05% BSA
Concentration 0.5mg/mL
Conjugate Unlabeled
Gene Accession No. NP_001026850.1
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 864